T2 Triol standard solution
Supplier: Apollo Scientific
About this item
Analytical reference standard solution in acetonitrile.
Caution: Expires in 12 months.
2 Options Available Below
Product Details & Documents
Analytical reference standard solution in acetonitrile.
Caution: Expires in 12 months.
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- APOSBIMS113-1ML
- APOSBIMS113-5ML
- Description:
- T2 Triol
- T2 Triol
- CAS No.:
- 34114-98-2
- 34114-98-2
- Formula:
- C₂₀H₃₀O₇
- C₂₀H₃₀O₇
- Flash point:
- 2 °C
- 2 °C
- Storage temperature:
- Freezer
- Freezer
Specifications
Product Family Options
Product Information
- ConcentrationPack typePkAvailabilityPrice
- APOSBIMS113-1MLT2 TriolSpecifications:
More
Specifications
Cat. No.APOSBIMS113-1MLDescriptionT2 TriolCAS No.34114-98-2FormulaC₂₀H₃₀O₇Flash point2 °CStorage temperatureFreezer - APOSBIMS113-5MLT2 TriolSpecifications:
More
Specifications
Cat. No.APOSBIMS113-5MLDescriptionT2 TriolCAS No.34114-98-2FormulaC₂₀H₃₀O₇Flash point2 °CStorage temperatureFreezer



