Hose connectors, b.safe
Supplier: Bohlender
Certificates
About this item
For the connection of elastic tubing (e.g. Viton, Tygon, silicone) on b.safe waste caps.
- Excellent chemical resistance
- Available for a wide range of hose diameters
- The conical shape gets the connection tight and leak-free
b.safe hose connectors are the flexible choice to customise your b.safe waste cap. Made of PP or PTFE and available in numerous diameters you will find the exact one to fit your waste hose.
Frequently Bought Together