
Tris(1H,1H,5H-octafluoropentyl)phosphate 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:355-86-2
:
Formula:C₁₅H₉F₂₄O₄P
MDL Number:MFCD00054654
:740.17 g/mol
Boiling Pt:130 °C/0.1mmHg
Flash Pt:>100 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together