
2,4,6-Tris(heptafluoropropyl)-1,3,5-triazine
Catalog # APOSPC7890-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:585.12 g/mol
Boiling Pt:165 °C
Density:1.716 g/cm³ (25 °C)
:GHS07
Storage Temperature:Ambient
CAS Number:915-76-4
Formula:C₁₂F₂₁N₃
MDL Number:MFCD00042438