Order Entry
Hungary
ContactUsLinkComponent
Comply™ Hydrogen Peroxide Chemical Indicator 1248
Comply™ Hydrogen Peroxide Chemical Indicator 1248
Catalog # MMMEM1248
Supplier:  3M MEDICAL
undefined
Comply™ Hydrogen Peroxide Chemical Indicator 1248
Catalog # MMMEM1248
Supplier:  3M MEDICAL
Specifications
  • Width:
    100 mm
  • Description:
    Hydrogen peroxide chemical indicator
  • Length:
    200 mm
  • Pk:
    250
Specifications
About this item
Internal pack monitor for use with Vaporised Hydrogen Peroxide/Gas Plasma sterilisation processes.
  • For use in STERRAD® sterilisation processes
  • Can be validated to other vaporised hydrogen peroxide cycles
  • Blue to pink colour change
Internal pack monitor for use in STERRAD® sterilisation processes (Models 100, 100S, NX Standard and Advanced cycles, and 100NX Standard, Flex, Express, Duo cycles). Indicator changes from blue to pink when exposed to Vaporised Hydrogen Peroxide. Indicator colour does not fade when exposed to light.