Order Entry
Hungary
ContactUsLinkComponent
Sterilisation wraps, sequential, KC100 KIMGUARD™
Sterilisation wraps, sequential, KC100 KIMGUARD™
Catalog # KIMB10712
Supplier:  KIMBERLY CLARK
   Waste Reducing
Sterilisation wraps, sequential, KC100 KIMGUARD™
Catalog # KIMB10712
Supplier:  KIMBERLY CLARK
   Waste Reducing
Specifications
  • Colour:
    Light blue
  • Description:
    Sterilisation wrap, Kimguard® KC300
  • Dimensions:
    305×305 mm
  • Seq:
    01
  • Environmentally Preferable:
    Waste Reducing
  • Pk:
    1.000
Specifications
About this item
Available in six strengths, from KC100 to KC600, to offer precisely the right protection for your needs
KIMGUARD™ KC100 Sterilization Wraps are the first of six strength levels. Printed with its name, size, and lot code.
Choosing is easy; the higher the number, the stronger the wrap. Sequential wrap. Used to package linen towels, packs, and gowns, light basin sets, and single instruments for sterilization. Low-lint fabric delivers barrier and strength properties that help prevent contamination after sterilization. Steam and EO compatible. Naturally latex-free. Non sterile. Multiple sizes.
Ordering information: Case count indicates the number of sheets included.