Order Entry
Hungary
ContactUsLinkComponent
VWR® Nitrile Light Ocean Blue, Nitrile Gloves
VWR® Nitrile Light Ocean Blue, Nitrile Gloves
Supplier:  VWR Collection
Certificates
undefined
VWR® Nitrile Light Ocean Blue, Nitrile Gloves
Supplier:  VWR Collection
Specifications
  • Catalog No:
  • 112-6069
  • 112-6067
  • 112-6070
  • Ambidextrous / hand-specific:
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Disposable / reusable:
  • Disposable
  • Disposable
  • Disposable
  • Latex / latex-free:
  • Latex-free
  • Latex-free
  • Latex-free
  • Powdered / powder-free:
  • Powder-free
  • Powder-free
  • Powder-free
  • Material:
  • Nitrile
  • Nitrile
  • Nitrile
  • Finish:
  • Textured fingertips
  • Textured fingertips
  • Textured fingertips
  • Cuff style:
  • Rolled
  • Rolled
  • Rolled
  • Finger thickness:
  • 0,10 mm, single wall
  • 0,10 mm, single wall
  • 0,10 mm, single wall
  • Palm thickness:
  • 0,08 mm, single wall
  • 0,08 mm, single wall
  • 0,08 mm, single wall
  • EN norm:
  • EN ISO 374-1 Type B (KTP), EN ISO 374-5 VIRUS
  • EN ISO 374-1 Type B (KTP), EN ISO 374-5 VIRUS
  • EN ISO 374-1 Type B (KTP), EN ISO 374-5 VIRUS
  • Food contact approved:
  • +
  • +
  • +