Order Entry
Hungary
ContactUsLinkComponent
Sodium acetate trihydrate 99.0-101.0% (dry basis), crystals USP, Multi-Compendial, J.T.Baker®
Sodium acetate trihydrate 99.0-101.0% (dry basis), crystals USP, Multi-Compendial, J.T.Baker®
Catalog # CH06-07
Supplier:  Avantor
CAS Number:  
undefined
Sodium acetate trihydrate 99.0-101.0% (dry basis), crystals USP, Multi-Compendial, J.T.Baker®
Catalog # CH06-07
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    PE pail, screw top
  • Pk:
    12 kg
  • Seq:
    4187
  • Pk Info:
    1×12 kg

Specifications

Specification Test Results

  • USP - Assay (CH₃COONa) (dried basis):
    99.0 - 101.0 %
  • USP - Calcium and Magnesium:
    Passes Test
  • USP - Chloride (Cl):
    ≤ 0.035 %
  • USP - Endotoxin Concentration (EU/g):
    ≤ 2.5
  • USP - Identification:
    Passes Test
  • USP - Insoluble Matter:
    ≤ 0.05 %
  • USP - Loss on Drying:
    38.0 - 41.0 %
  • USP - pH of 5% Solution at 25°C:
    7.5 - 9.2
  • USP - Potassium (K):
    Passes Test
  • USP - Sulfate (SO₄):
    ≤ 0.005 %
  • EP/BP - Assay (CH₃COONa):
    99.0 - 101.0 %
  • EP/BP - Identification A:
    Passes Test
  • EP/BP - Identification B:
    Passes Test
  • EP/BP - Identification C:
    Passes Test
  • EP/BP - Appearance of Solution:
    Passes Test
  • EP/BP - pH (1 in 20):
    7.5 - 9.0
  • EP/BP - Reducing Substances:
    Passes Test
  • EP/BP - Chloride (Cl):
    ≤ 200 ppm
  • EP/BP - Sulfate (SO₄):
    ≤ 200 ppm
  • EP/BP - Calcium and Magnesium:
    ≤ 50 ppm
  • EP/BP - Iron (Fe):
    ≤ 10 ppm
  • EP/BP - Loss on Drying:
    39.0 - 40.5 %
  • JP - Assay (CH₃COONa):
    99.5 - 101.0 %
  • JP - Identification:
    Passes Test
  • JP - Clarity and Color of Solution:
    Passes Test
  • JP - Acidity or Alkalinity:
    Passes Test
  • JP - Chloride (Cl):
    ≤ 0.011 %
  • JP - Sulfate (SO₄):
    ≤ 0.017 %
  • JP - Heavy Metals (as Pb):
    ≤ 10 ppm
  • JP - Calcium and Magnesium:
    Passes Test
  • JP - Arsenic (As):
    ≤ 2 ppm
  • JP - Substances Reducing Permanganate:
    Passes Test
  • JP - Loss on Drying:
    39.0 - 40.5 %
  • FCC - Alkalinity:
    ≤ 0.05 %
  • FCC - Assay (CH₃COONa) (dried basis):
    99.0 - 101.0 %
  • FCC - Identification A:
    Passes Test
  • FCC - Identification B:
    Passes Test
  • FCC - Lead (Pb):
    ≤ 2 mg/kg
  • FCC - Loss on Drying:
    38.0 - 41.0 %
  • FCC - Potassium Compounds:
    Passes Test
  • ChP - Assay (dried basis):
    ≥ 99.0 %
  • ChP - Identification:
    Passes Test
  • ChP - Alkalinity (pH):
    7.5 - 9.2
  • ChP - Chloride:
    ≤ 0.035 %
  • ChP - Sulfate:
    ≤ 0.005 %
  • ChP - Loss on Drying:
    38.0 - 41.0 %
  • ChP - Insoluble matter:
    ≤ 0.05 %
  • ChP - Calcium and Magnesium:
    Passes Test
  • ChP - Potassium:
    Passes Test
  • ChP - Iron (Fe):
    ≤ 0.001 %
  • ChP - Heavy metals:
    ≤ 0.001
  • ChP - Arsenic:
    ≤ 0.0003 %
  • ChP - Endotoxin Concentration, EU/g:
    ≤ 2.5

Specification Test Results

About this item

 :
 :136.08 g/mol
 :Management of Change (MOC) category = R
Flash Pt:250.0 °C
CAS Number:6131-90-4
Formula:H₃CCOONa·3H₂O
MDL Number:MFCD00071557