Order Entry
Hungary
ContactUsLinkComponent
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) USP, Multi-Compendial, J.T.Baker®
Irritant
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) USP, Multi-Compendial, J.T.Baker®
Catalog # 4101-09
Supplier:  Avantor
CAS Number:  
Irritant
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) USP, Multi-Compendial, J.T.Baker®
Catalog # 4101-09
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Drum with poly lockband
  • Pk:
    50 kg
  • Seq:
    4418

Specifications

Specification Test Results

  • USP - Assay (C₄H₁₁NO₃):
    99.0 - 101.0 %
  • USP - Identification A:
    Passes Test
  • USP - Identification B:
    Passes Test
  • USP - Identification C:
    Passes Test
  • USP - pH (1 in 20):
    10.0 - 11.5
  • USP - Residue on Ignition:
    ≤0.1 %
  • USP - Loss on Drying:
    ≤0.5 %
  • USP - Melting Range(Lower):
    168 - 172 °C
  • USP - Melting Range(Upper):
    168 - 172 °C
  • EP/BP - Assay (C₄H₁₁NO₃) (dried basis):
    99.0 - 100.5 %
  • EP/BP - Identification B:
    Passes Test
  • EP/BP - Identification C:
    Passes Test
  • EP/BP - Appearance of Solution:
    Passes Test
  • EP/BP - pH:
    10.0 - 11.5
  • EP/BP - Related Substances:
    ≤1.0 %
  • EP/BP - Chloride (Cl):
    ≤100 ppm
  • EP/BP - Iron (Fe):
    ≤10 ppm
  • EP/BP - Loss on Drying at 105°C:
    ≤0.5 %
  • EP/BP - Ash (sulfated):
    ≤0.1 %
  • JPC 1997 - Assay (C₄H₁₁NO₃) (dried basis):
    ≥99.0 %
  • JPC 1997 - Identification A:
    Passes Test
  • JPC 1997 - Identification B:
    Passes Test
  • JPC 1997 - pH:
    10.3 - 10.7
  • JPC 1997 - Melting Point:
    168 - 172 °C
  • JP - Clarity and Color of Solution:
    Passes Test
  • JPC 1997 - Heavy Metals (as Pb):
    ≤8 ppm
  • JPC 1997 - Arsenic (As):
    ≤1.6 ppm
  • JPC 1997 - Loss on Drying:
    ≤0.5 %
  • JPC 1997 - Residue on Ignition:
    ≤0.10 %
  • Assay (C₄H₁₁NO₃) (dried basis):
    ≥99.9 %
  • Absorbance of a 1M Solution - 260 nm:
    ≤0.06
  • Absorbance of a 1M Solution - 280 nm:
    ≤0.06
  • Absorbance of a 1M Solution - 400 nm:
    ≤0.01
  • Endotoxin Concentration (EU/g):
    ≤2.5
  • Iron (Fe):
    ≤1 ppm
  • Lead (Pb):
    ≤1 ppm
  • Water (H₂O)(by Karl Fischer titrn):
    ≤0.5 %
  • Enzyme Activity - DNase Activity:
    Passes Test
  • Enzyme Activity - RNase Activity:
    Passes Test
  • Enzyme Activity - Protease Activity:
    Passes Test
  • Trace Impurities - Arsenic (As):
    ≤1 ppm
  • Trace Impurities - Aluminum (Al):
    ≤5.000 ppm
  • Trace Impurities - Barium (Ba):
    ≤5 ppm
  • Trace Impurities - Bismuth (Bi):
    ≤5 ppm
  • Trace Impurities - Calcium (Ca):
    ≤10.0 ppm
  • Trace Impurities - Cadmium (Cd):
    ≤5.000 ppm
  • Trace Impurities - Chromium (CR):
    ≤5 ppm
  • Trace Impurities - Cobalt (Co):
    ≤5.000 ppm
  • Trace Impurities - Copper (Cu):
    ≤5 ppm
  • Trace Impurities - Lithium (Li):
    ≤5 ppm
  • Trace Impurities - Magnesium (Mg):
    ≤5 ppm
  • Trace Impurities - Mercury (Hg):
    ≤5 ppm
  • Trace Impurities - Molybdenum (Mo):
    ≤5 ppm
  • Trace Impurities - Nickel (Ni):
    ≤5.000 ppm
  • Trace Impurities - Potassium (K):
    ≤50 ppm
  • Trace Impurities - Sodium (Na):
    ≤50.000 ppm
  • Trace Impurities - Strontium (Sr):
    ≤5 ppm
  • Trace Impurities - Vanadium (V):
    ≤5.000 ppm
  • Trace Impurities - Zinc (Zn):
    ≤5.000 ppm

Specification Test Results

About this item

 :
 :121.14 g/mol
 :GHS07
CAS Number:77-86-1
Formula:NH₂C(CH₂OH)₃
MDL Number:MFCD00004679