Order Entry
Hungary
ContactUsLinkComponent
Instrument check standard 3 for ICP
Instrument check standard 3 for ICP
Catalog # HONCE2109-100ML
CAS Number:  
Instrument check standard 3 for ICP
Catalog # HONCE2109-100ML
CAS Number:  

Specifications

  • Matrix:
    2% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Instrument Check Standard 3

Specifications

About this item

Certified Reference Material (CRM).

200 mg/L each of Ca, Fe, K, Mg, Na in HNO₃ 2%