Order Entry
Hungary
ContactUsLinkComponent
 
Avantor® Hichrom HI-5 MS Xil, GC Columns
 :  Avantor
 
undefined
Avantor® Hichrom HI-5 MS Xil, GC Columns
 :  Avantor
 

 

  • Catalog No:
  • HI19-25-050-105
  • HI19-18-040-40
  • HI19-25-050-60
  • HI19-25-050-50
  • HI19-25-025-60
  • HI19-18-020-40
  • HI19-32-100-105
  • HI19-25-025-30
  • HI19-18-020-10
  • HI19-32-100-50
  • HI19-32-100-30
  • HI19-32-150-30
  • HI19-32-150-15
  • HI19-32-150-105
  • HI19-18-040-10
  • HI19-18-018-60
  • HI19-25-025-105
  • HI19-25-025-10
  • HI19-25-010-60
  • HI19-32-025-30
  • HI19-20-100-50
  • HI19-20-100-40
  • HI19-53-150-50
  • HI19-25-025-60G10
  • HI19-18-018-20
  • HI19-32-050-105
  • HI19-25-025-60G5
  • HI19-25-025-30G5
  • HI19-32-025-50
  • HI19-25-050-30G5
  • HI19-32-050-60
  • HI19-15-015-10
  • HI19-32-025-30G5
  • HI19-32-050-50
  • HI19-18-018-50
  • HI19-25-050-15G10
  • HI19-10-040-5
  • HI19-18-018-40
  • HI19-25-050-30G10
  • HI19-32-050-15
  • HI19-15-015-5
  • HI19-32-100-30G5
  • HI19-15-015-30
  • HI19-53-100-30G5
  • HI19-15-015-20
  • HI19-20-040-60
  • HI19-25-010-30G5
  • HI19-20-033-30
  • HI19-25-025-15G5
  • HI19-53-050-50
  • HI19-20-020-60
  • HI19-53-100-50
  • HI19-20-040-10
  • HI19-32-010-105
  • HI19-25-025-30G10
  • HI19-25-100-60
  • HI19-53-100-15
  • HI19-05-005-5
  • HI19-32-010-60
  • HI19-05-005-2.5
  • HI19-05-005-2
  • HI19-10-020-10
  • HI19-10-010-5
  • HI19-32-025-15
  • HI19-32-025-105
  • HI19-10-010-10
  • HI19-20-020-50
  • HI19-20-020-20
  • HI19-53-010-10
  • HI19-25-100-15
  • HI19-18-040-50
  • HI19-25-100-105
  • HI19-53-025-50
  • HI19-53-025-15
  • HI19-53-010-30
  • HI19-20-010-40
  • HI19-53-050-15
  • HI19-53-050-10
  • Phase name:
  • HI-5 MS Xil
  • HI-5 MS Xil
  • HI-5 MS Xil
  • HI-5 MS Xil
  • HI-5 MS Xil
  • HI-5 MS Xil
  • HI-5 MS Xil
  • HI-5 MS Xil
  • HI-5 MS Xil
  • HI-5 MS Xil
  • HI-5 MS Xil
  • HI-5 MS Xil
  • HI-5 MS Xil
  • HI-5 MS Xil
  • HI-5 MS Xil
  • HI-5 MS Xil
  • HI-5 MS Xil
  • HI-5 MS Xil
  • HI-5 MS Xil
  • HI-5 MS Xil
  • HI-5 MS Xil
  • HI-5 MS Xil
  • HI-5 MS Xil
  • HI-5 MS Xil
  • HI-5 MS Xil
  • HI-5 MS Xil
  • HI-5 MS Xil
  • HI-5 MS Xil
  • HI-5 MS Xil
  • HI-5 MS Xil
  • HI-5 MS Xil
  • HI-5 MS Xil
  • HI-5 MS Xil
  • HI-5 MS Xil
  • HI-5 MS Xil
  • HI-5 MS Xil
  • HI-5 MS Xil
  • HI-5 MS Xil
  • HI-5 MS Xil
  • HI-5 MS Xil
  • HI-5 MS Xil
  • HI-5 MS Xil
  • HI-5 MS Xil
  • HI-5 MS Xil
  • HI-5 MS Xil
  • HI-5 MS Xil
  • HI-5 MS Xil
  • HI-5 MS Xil
  • HI-5 MS Xil
  • HI-5 MS Xil
  • HI-5 MS Xil
  • HI-5 MS Xil
  • HI-5 MS Xil
  • HI-5 MS Xil
  • HI-5 MS Xil
  • HI-5 MS Xil
  • HI-5 MS Xil
  • HI-5 MS Xil
  • HI-5 MS Xil
  • HI-5 MS Xil
  • HI-5 MS Xil
  • HI-5 MS Xil
  • HI-5 MS Xil
  • HI-5 MS Xil
  • HI-5 MS Xil
  • HI-5 MS Xil
  • HI-5 MS Xil
  • HI-5 MS Xil
  • HI-5 MS Xil
  • HI-5 MS Xil
  • HI-5 MS Xil
  • HI-5 MS Xil
  • HI-5 MS Xil
  • HI-5 MS Xil
  • HI-5 MS Xil
  • HI-5 MS Xil
  • HI-5 MS Xil
  • HI-5 MS Xil
  • Phase composition:
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Silphenylene Methyl Polysiloxane - low bleeding
  • Polarity:
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Crossbond:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Temperature limit:
  • 360 °C
  • 360 °C
  • 360 °C
  • 360 °C
  • 360 °C
  • 360 °C
  • 360 °C
  • 360 °C
  • 360 °C
  • 360 °C
  • 360 °C
  • 360 °C
  • 360 °C
  • 360 °C
  • 360 °C
  • 360 °C
  • 360 °C
  • 360 °C
  • 360 °C
  • 360 °C
  • 360 °C
  • 360 °C
  • 360 °C
  • 360 °C
  • 360 °C
  • 360 °C
  • 360 °C
  • 360 °C
  • 360 °C
  • 360 °C
  • 360 °C
  • 360 °C
  • 360 °C
  • 360 °C
  • 360 °C
  • 360 °C
  • 360 °C
  • 360 °C
  • 360 °C
  • 360 °C
  • 360 °C
  • 360 °C
  • 360 °C
  • 360 °C
  • 360 °C
  • 360 °C
  • 360 °C
  • 360 °C
  • 360 °C
  • 360 °C
  • 360 °C
  • 360 °C
  • 360 °C
  • 360 °C
  • 360 °C
  • 360 °C
  • 360 °C
  • 360 °C
  • 360 °C
  • 360 °C
  • 360 °C
  • 360 °C
  • 360 °C
  • 360 °C
  • 360 °C
  • 360 °C
  • 360 °C
  • 360 °C
  • 360 °C
  • 360 °C
  • 360 °C
  • 360 °C
  • 360 °C
  • 360 °C
  • 360 °C
  • 360 °C
  • 360 °C
  • 360 °C
  • USP code:
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41

 

 

Frequently Bought Together