
5-Hydroxysophthalic acid
Catalog # MOLE24242615-100G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:182.13 g/mol
Boiling Pt:386 °C (1013 hPa)
Melting Pt:298…301 °C
EINECS:210-565-7
:GHS07
CAS Number:618-83-7
Formula:C₈H₆O₅
MDL Number:MFCD00002515