

1-Cetylpyridinium chloride monohydrate
Catalog # MOLE29862237-500G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
- Seq:3645
Specifications
About this item
:
:358.01 g/mol
Merck Index:13,02041
Melting Pt:77…83 °C
Storage Temperature:Ambient
EINECS:204-593-9
UN:2811
ADR:6.1,I
:GHS06
:GHS09
CAS Number:6004-24-6
Formula:C₂₁H₃₈ClN·1H₂O
MDL Number:MFCD00149977