Order Entry
Hungary
ContactUsLinkComponent
Sodium nitroprusside dihydrate 99.0-102.0%, AnalaR NORMAPUR® analytical reagent
Toxic
Sodium nitroprusside dihydrate 99.0-102.0%, AnalaR NORMAPUR® analytical reagent
Catalog # 27966.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
Toxic
Sodium nitroprusside dihydrate 99.0-102.0%, AnalaR NORMAPUR® analytical reagent
Catalog # 27966.180
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Assay:
    99.0 - 102.0 %
  • Hexacyanoferrate (II) (as Fe(CN)6):
    Max. 0.02 %
  • Hexacyanoferrate (III) (as Fe(CN)6):
    Max. 0.02 %
  • Insolubility in methanol:
    Max. 0.5 %
  • Insoluble matter:
    Max. 100 ppm
  • Cl (Chloride):
    Max. 100 ppm
  • SO4 (Sulphate):
    Max. 100 ppm

Specification Test Results

About this item

 :
Merck Index:13,08722
Density:1.72 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:1588
ADR:6.1,III
 :297.95 g/mol
EINECS:238-373-9
 :GHS06
CAS Number:13755-38-9
Formula:Na₂[Fe(CN)₅NO]·2H₂O
MDL Number:MFCD00149192