

Acetic acid ≥99.8% (by HPLC), HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:64-19-7
:
Formula:H₃CCOOH
MDL Number:MFCD00036152
:60.05 g/mol
UN:2789
ADR:8,II
:GHS02
:GHS05
Limited Time Sale
Select items below are currently on sale
Specification Test Results
- Assay (on anhydrous substance) :Min. 99.8 %
- Evaporation residue :Max. 0.0001 %
- Water :Max. 0.1 %
- Transmittance (252 nm) :Min.10 %
- Transmittance (260 nm) :Min. 50 %
- Transmittance (270 nm) :Min. 80 %
- Transmittance (280 nm) :Min. 95 %
- Transmittance (300 nm) :Min. 99 %
Specification Test Results
Related Information
Frequently Bought Together