Order Entry
Hungary
ContactUsLinkComponent
 
VWR®, Clamp Holder
  444-0917
VWR®, Clamp Holder
  444-0917

 

  • Description:
    Regular clamp holder, 0 - 18 mm grip size
  • Material:
    Nickel-plated zinc
  • Pk:
    1

 

 

This regular clamp holder is made of nickel-plated zinc construction.

  • Ideal for holding clamps to lab-frames
  • Use wherever clamping at 90° is required