Test sieves, 305×40 mm
Supplier: Retsch GmbH
Certificates
About this item
Sieves made from 304 stainless steel are individually laser engraved with clear and precise labelling for full traceability.
- Sieve bodies are made in one piece and the absence of hollow spaces prevents cross-contamination
- High corrosion resistance and easy to clean
- Maximum stability and optimal sealing of the sieve stack, 15% less weight than traditional sieves
Sieve meshes, frames and labeling comply with standards. The sieves are tested five times and are delivered with test report.
Certifications: An individual inspection certificate for test materials monitoring according to ASTM E 11 is available
Ordering information: Sieves and accessories of other dimensions are available on request. Also available with perforated plate, round or square.
Frequently Bought Together