
Sodium hydroxide solution ≥32% in water for nitrogen analysis (max. 0.0001% N)
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:1310-73-2
:
Formula:NaOH
MDL Number:MFCD00003548
:40 g/mol
Boiling Pt:120 °C
Melting Pt:9…10 °C
Density:1.350 g/cm³
Storage Temperature:Ambient
UN:1824
ADR:8,II
:GHS05
Specification Test Results
- Assay:31.5 - 32.5 %
- Assay of Na₂CO₃:Max. 0.5 %
- Aluminium (Al):Max. 0.0005 %
- Iron (Fe):Max. 0.0005 %
- Heavy metals (as Pb):Max. 0.0005 %
- Chloride (Cl):Max. 0.003 %
- Phosphate (PO₄):Max. 0.0005 %
- Silicate (as SiO₂):Max. 0.001 %
- Sulfate (SO₄):Max. 0.0005 %
- Total N:Max. 0.0001 %
Specification Test Results
Frequently Bought Together