Order Entry
Hungary
ContactUsLinkComponent
 
 
 
Lithium dodecyl sulphate 98+% (by acidimetry)
 
 
 
Lithium dodecyl sulphate 98+% (by acidimetry)

 

  • Form:
    White crystalline powder
  • Assay (unspecified):
    ≥98% (acidity)
  • Assay (unspecified) Absorbance (230 nm, 3%, 1 cm):
    ≤ 0.2
  • Assay (unspecified) Absorbance (280 nm, 3%, 1 cm):
    ≤ 0.1
  • Assay (GC) (C12):
    ≥98%
  • Chloride content:
    ≤ 0.01%
  • Loss on Drying:
    ≤ 1%
  • Trace Metal: Copper (Cu):
    ≤ 5 ppm
  • Trace Metal: Iron (Fe):
    ≤ 1 ppm
  • Trace Metal: Lead (Pb):
    ≤ 5 ppm

 

Frequently Bought Together