Acid special cleaner, TICKOPUR R 27
Supplier: BANDELIN-ELECTRONIC
Certificates
About this item
Special cleaner based on phosphoric acid, for decalcification and rust removal with corrosion protection.
Frequently Bought Together
Special cleaner based on phosphoric acid, for decalcification and rust removal with corrosion protection.