
Tricyclohexylphosphine
Catalog # MOLE18931439-5G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:280.43 g/mol
Boiling Pt:132…134 °C (0.02 mmHg)
Boiling Pt:132…134 °C (0,02 mmHg)
Melting Pt:78 °C
Density:0,9 g/cm³ (20 °C)
Storage Temperature:Freezer
EINECS:220-069-2
:GHS07
CAS Number:2622-14-2
Formula:C₁₈H₃₃P
MDL Number:MFCD00003853