ω-Bromo-4-chloroacetophenone
Catalog # MOLE23463177-100G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:233.49 g/mol
Merck Index:13,02172
Boiling Pt:278 °C (1013 hPa)
Melting Pt:110 °C
EINECS:208-631-5
UN:3261
ADR:8,III
CAS Number:536-38-9
Formula:C₈H₆BrClO
MDL Number:MFCD00000625