Order Entry
Hungary
ContactUsLinkComponent
 
 
 
 
 
n-Pentane ≥99.0%, PESTINORM® for capillary GC analysis
 
undefined
 
 
 
 
n-Pentane ≥99.0%, PESTINORM® for capillary GC analysis
 
 
 

 

  • Assay (on anhydrous substance):
    Min. 99.0 %
  • Acidity:
    Max. 0.0005 meq/g
  • Evaporation residue (100°C):
    Max. 0.0005 %
  • Water:
    Max. 0.01 %
  • Organic residue (as Octanol) (GC/FID):
    Max. 10 ng/l
  • Halogenated residue (as Lindane)(GC/ECD):
    Max. 5 ng/l

 

Frequently Bought Together