Order Entry
Hungary
ContactUsLinkComponent
 
Flammable
Irritant
Triethyl orthopropionate
Catalog # MOLE45664077-100G
Supplier:  Molekula
CAS Number:  
Flammable
Irritant
Triethyl orthopropionate
Catalog # MOLE45664077-100G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :176.26 g/mol
Boiling Pt:155 °C (1013 hPa)
Melting Pt:< 0 °C
Density:0.8876 g/cm³ (20 °C)
Flash Pt:39 °C
Storage Temperature:Ambient
EINECS:204-108-0
UN:3272
ADR:3,III
 :GHS02
 :GHS07
CAS Number:115-80-0
Formula:C₉H₂₀O₃
MDL Number:MFCD00009226