Order Entry
Hungary
ContactUsLinkComponent
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) ≥99.9% buffer substance, for analysis and biochemical purposes
Irritant
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) ≥99.9% buffer substance, for analysis and biochemical purposes
Supplier:  VWR Chemicals
Certificates
undefined
Irritant
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) ≥99.9% buffer substance, for analysis and biochemical purposes
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay (calculated on dried substance):
    Min. 99.9 %
  • Colouration (20 %; water):
    Max. 20 APHA
  • Melting point:
    Min. 170 °C
  • pH (20°C; 5 %):
    10.0 - 11.5
  • Heavy metals (as Pb):
    Max. 1 ppm
  • Insolubility in water:
    Max. 50 ppm
  • Water:
    Max. 0.2 %
  • Absorbance (260 nm) (1 mol/l):
    Max 0.05
  • Absorbance (280 nm) (1 mol/l):
    Max. 0.03
  • Absorbance (430 nm) (1 mol/l):
    Max. 0.004

Specification Test Results

Frequently Bought Together