Order Entry
Hungary
ContactUsLinkComponent
Zinc sulphate heptahydrate ≥99%, GPR RECTAPUR®
Irritant
Corrosive
Environment
Zinc sulphate heptahydrate ≥99%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
Irritant
Corrosive
Environment
Zinc sulphate heptahydrate ≥99%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 99 %
  • Cl (Chloride):
    Max. 0.03 %
  • Fe (Iron):
    Max. 20 ppm
  • Pb (Lead):
    Max. 100 ppm

Specification Test Results

Frequently Bought Together