Order Entry
Hungary
ContactUsLinkComponent
Sodium acetate trihydrate 99.5-101.0%, EMPROVE® ESSENTIAL Ph. Eur., BP, USP, JP, FCC, E262, extra pure, SAFC®
Sodium acetate trihydrate 99.5-101.0%, EMPROVE® ESSENTIAL Ph. Eur., BP, USP, JP, FCC, E262, extra pure, SAFC®
  1.06265.9027
 :  
undefined
Sodium acetate trihydrate 99.5-101.0%, EMPROVE® ESSENTIAL Ph. Eur., BP, USP, JP, FCC, E262, extra pure, SAFC®
  1.06265.9027
 :  

 

  • Pack type:
    PE sack, double
  • Pk:
    25 kg
  • Seq:
    4283

 

 

  • Assay (perchloric acid titration, calculated on dried substance):
    99.5 - 101.0 %
  • Identity:
    passes test
  • Appearance of solution:
    passes test
  • In water insoluble matter:
    ≤ 0.03 %
  • Alkalinity:
    ≤ 0.05 %
  • Acidity or alkalinity:
    passes test
  • pH-value (1 %, water):
    8.0 - 9.5
  • pH-value (5 %; water):
    7.5 - 9.0
  • Chloride (Cl):
    ≤ 0.005 %
  • Sulfate (SO₄):
    ≤ 0.0030 %
  • Heavy metals (as Pb):
    ≤ 0.0006 %
  • Al (Aluminium):
    ≤ 0.00002 %
  • As (Arsenic):
    ≤ 0.0002 %
  • Ca (Calcium):
    ≤ 0.002 %
  • Fe (Iron):
    ≤ 0.0010 %
  • Hg (Mercury):
    ≤ 0.0001 %
  • K (Potassium):
    ≤ 0.005 %
  • Mg (Magnesium):
    ≤ 0.001 %
  • Pb (Lead):
    ≤ 0.0002 %
  • Ca + Mg (calcium and magnesium):
    passes test
  • free acetic acid:
    ≤ 0.4 %
  • Other residual solvents (ICH Q3C):
    excluded by manufacturing process
  • formic acid, formate and other oxidizable impurities:
    ≤ 0.1000 %
  • Substances reducing potassium permanganate (as HCOOH):
    passes test
  • Loss on drying (130 °C):
    39.0 - 40.5 %

 

 

 :
 :136.08 g/mol
 :13,08642
 :120…123 °C (1013 hPa)
 :58 °C
 :1.4 g/cm³ (20 °C)
 :Ambient
 :204-823-8
 :6131-90-4
 :H₃CCOONa·3H₂O
 :MFCD00071557