
2,3,5-Triphenyltetrazolium chloride (TTC) ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:298-96-4
:
Formula:C₁₉H₁₅ClN₄
MDL Number:MFCD00011963
Merck Index:14,9744
Boiling Pt:∼360 °C (1013 hPa)
Melting Pt:∼ 250 °C (dec.)
:334.81 g/mol
Storage Temperature:Refrigerator
EINECS:206-071-6
:GHS07
Frequently Bought Together