Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 Ft
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Hungary
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Sodium hydroxide
Sodium hydroxide standardised solution 1.0 N in aqueous solution
Sodium hydroxide standardised solution 1.0 N in aqueous solution
Sodium hydroxide standardised solution 1.0 N in aqueous solution
35629.K2
:
Thermo Fisher Scientific
:
Sodium hydroxide standardised solution 1.0 N in aqueous solution
35629.K2
:
Thermo Fisher Scientific
:
Pack type:
Plastic bottle
Pk:
1 L
Seq:
5037
:
:
40 g/mol
:
1.04 g/cm³ (25 °C)
:
Ambient
:
215-185-5
:
3266
:
8,III
:
GHS05
:
1310-73-2
:
NaOH
:
MFCD00003548