Order Entry
Hungary
Orders LinkContactUsLinkComponent
cis-Platin

cis-Platin

Glass bottle for solids, 100 mg
Catalog # PROOICRS0258
cis-Platin
Catalog # PROOICRS0258
CAS Number:  15663-27-1
cis-Platin

Glass bottle for solids, 100 mg

CAS Number:  15663-27-1
Price
157 000 Ft
Log in for availability
1 * 100 mg
Product Details & Documents
Synonyms:  Platinol, cis-Diaminodichloroplatinum(II), cis-Diamminedichloroplatinum (II), cis-Dichlorodiammine platinum(II), Cisplatin, cis-Diamineplatinum(II) dichloride
  • CAS Number:15663-27-1
  • Formula:H₆Cl₂N₂Pt
  • MDL Number:MFCD00011623
  • Molecular Weight:300.05 g/mol
  • Merck Index:13,02341
  • Density:3.738 g/cm³ (20 °C)
  • EINECS:239-733-8
  • UN:3288
  • ADR:6.1,II
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg

Specifications