Chloroform-[D₁] 99.8 atom % D stabilised for NMR spectroscopy
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Trichloromethane-D1, Chloroform-[²H₁], Chloroform-D
CAS Number:865-49-6
:
Formula:CDCl₃
MDL Number:MFCD00000827
:120.38 g/mol
Boiling Pt:60.8 °C (1013 hPa)
Melting Pt:–64 °C
Density:1.5 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:212-742-4
ADR:6.1,III
:Stabilised with silver
UN:1888
:GHS06
:GHS08
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Water:≤0.02 % (Coulometric)
- D-Enrichment:≥99.75 %
Specification Test Results
Frequently Bought Together