Order Entry
Hungary
Orders LinkContactUsLinkComponent
Tin

Tin

Glass bottle for solids, 200 g
Catalog # PROONIST-741A
Tin
Catalog # PROONIST-741A
CAS Number:  7440-31-5
Tin

Glass bottle for solids, 200 g

CAS Number:  7440-31-5
Price
2 906 000 Ft
Log in for availability
1 * 200 g
Product Details & Documents
  • CAS Number:7440-31-5
  • Formula:Sn
  • MDL Number:MFCD00133862
  • Molecular Weight:118.71 g/mol
  • Merck Index:12,09587
  • Boiling Pt:2270 °C (1013 hPa)
  • Melting Pt:232 °C
  • Density:7.31 g/cm³ (25 °C)
  • Storage Temperature:Ambient
  • EINECS:231-141-8
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    200 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.