Order Entry
Hungary
ContactUsLinkComponent
Human recombinant VHR dual specificity phosphatase (from E. coli)
Human recombinant VHR dual specificity phosphatase (from E. coli)
Catalog # ENZOBMLSE3330050
Supplier:  Enzo Life Sciences
CAS Number:  
Human recombinant VHR dual specificity phosphatase (from E. coli)
Catalog # ENZOBMLSE3330050
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Seq:
    02
  • Enzyme name:
    VHR dual specificity phosphatase

Specifications

About this item

Produced in E. coli.