Order Entry
Hungary
ContactUsLinkComponent
 
Mouse recombinant caspase-11 (from E. coli)
Catalog # ENZOBMLSE1555000
Supplier:  Enzo Life Sciences
CAS Number:  
Mouse recombinant caspase-11 (from E. coli)
Catalog # ENZOBMLSE1555000
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    1 Vial
  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Seq:
    02
  • Enzyme name:
    Caspase-11
  • Enzyme activity:
    5000 U

Specifications

About this item

Produced in E. coli. Mouse caspase-11 (aa 81-373).