Order Entry
Hungary
ContactUsLinkComponent
Human recombinant Guanylyl cyclase alpha1beta1 (soluble) (from Sf9 cells)
Human recombinant Guanylyl cyclase alpha1beta1 (soluble) (from Sf9 cells)
Katalógus# ENZOALX201180R050
Supplier:  Enzo Life Sciences
CAS Number:  
Human recombinant Guanylyl cyclase alpha1beta1 (soluble) (from Sf9 cells)
Katalógus# ENZOALX201180R050
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    50 µl
  • Enzyme type:
    Recombinant
  • Source:
    Insect cells (Sf9)
  • Species:
    Human
  • Seq:
    02
  • Enzyme name:
    Guanylyl cyclase alpha1beta1 (soluble)
  • Concentration:
    ≥1,5 mg/ml (total protein).
  • Formulation:
    Liquid. Enriched fraction in 50 mM TEA, pH 7,4 containing 1 mM EDTA/EGTA, 5 mM GSH and 30% glycerol.

Specifications

About this item

Produced in Sf9 cells. Fused at the C-terminus to a His-tag.