Human recombinant Guanylyl cyclase alpha1beta1 (soluble) (from Sf9 cells)
Catalog # ENZOALX201177C010
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:10 µG
- Enzyme type:Recombinant
- Source:Insect cells (Sf9)
- Species:Human
- Seq:02
- Enzyme name:Guanylyl cyclase alpha1beta1 (soluble)
- Purity:≥90% (SDS-PAGE)
- Concentration:0,4 mg/ml
- Formulation:In 40 mM TEA, pH 7,4 containing 2 mM DTT, 1 mM MgCl₂ and 30% glycerol.
Specifications
About this item
Produced in Sf9 cells. Fused at the C-terminus to a His-tag.