Order Entry
Hungary
ContactUsLinkComponent
Human recombinant Caspase-6 (from E. coli)
Human recombinant Caspase-6 (from E. coli)
Supplier:  Enzo Life Sciences
Certificates
Human recombinant Caspase-6 (from E. coli)
Supplier:  Enzo Life Sciences

Specifications

  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Form:
    Lyophilised
  • Reconstitution instructions:
    Reconstitute to 1 U/µl with PBS containing 15% glycerol.
  • Enzyme name:
    Caspase-6
  • Purity:
    ≥95% (SDS-PAGE)

Specifications

Frequently Bought Together