Human recombinant Caspase-3 (from E. coli)
Supplier: Enzo Life Sciences
Certificates
About this item
Produced in E. coli.
Specifications
- Enzyme type:Recombinant
- Source:E. coli
- Species:Human
- Form:Lyophilised
- Reconstitution instructions:Reconstitute to 1 U/µl with PBS containing 15% glycerol.
- Enzyme name:Caspase-3
Specifications
Frequently Bought Together