Human recombinant Cathepsin S
Catalog # ENZOBMLSE4530010
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:10 µG
- Protein/peptide type:Recombinant
- Species:Human
- Seq:02
- Protein/peptide name:Cathepsin S
Specifications
About this item
Highly active
Produced in insect cells. Recombinant active cathepsin S cloned from human cDNA as full length proenzyme, auto-activated by incubation in pH 4,5 acetate buffer, and purified.