CDK5 substrate
Catalog # ENZOBMLP2630001
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:1 mg
- Seq:02
- Protein/peptide name:CDK5 Substrate
- Purity:>95% (HPLC)
- Formulation:Lyophilized
Specifications
About this item
This peptide, PKTPKKAKKL, is derived from histone H1 and is a substrate for CDK5.