Order Entry
Hungary
ContactUsLinkComponent
Shp 1 and 2 substrate
Shp 1 and 2 substrate
Catalog # ENZOBMLP1570001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Shp 1 and 2 substrate
Catalog # ENZOBMLP1570001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Seq:
    02
  • Protein/peptide name:
    Shp 1 and 2 substrate

Specifications

About this item

This peptide sequence, EFpYAEVGRSPPDPAK, was selected by phage display analysis for optimal Shp-1 and Shp-2 phosphatase activities.