Ac-YVAD-AFC
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
Substrate for caspase-1 and -4
Fluorogenic substrate for caspase-1 and -4.
Specifications
- Protein/peptide name:Ac-YVAD-AFC
- Purity:>99% (HPLC)