Human recombinant CARM1 (from Insect cells (Sf9)), HIS Tag
Catalog # ENZOALX201283C002
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:2 µG
- Protein/peptide type:Recombinant
- Source:Insect cells (Sf9)
- Species:Human
- Seq:02
- Tag sequence:HIS Tag
- Protein/peptide name:CARM1
- Purity:>90% (SDS-PAGE)
- Concentration:0,5 mg/ml
- Formulation:Liquid. In 50 mM TRIS-HCl, pH 7,5, containing 100 mM sodium chloride, 0,2% NP-40, 50 mM imidazole and 10% glycerol.
Specifications
About this item
Produced in SF21 cells. Fused to a His-tag.