Order Entry
Hungary
ContactUsLinkComponent
Fluor-de-Lys® SIRT1 fluorometric drug discovery assay kit
Fluor-de-Lys® SIRT1 fluorometric drug discovery assay kit
Catalog # ENZOBMLAK5550001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Fluor-de-Lys® SIRT1 fluorometric drug discovery assay kit
Catalog # ENZOBMLAK5550001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Description:
    Fluor-de-Lys® SIRT1 fluorometric drug discovery assay kit
  • Seq:
    02
  • Storage conditions:
    –80 °C (long term)
  • Application:
    Fluorescent detection, HTS activity assay, cell-based assays
  • Size:
    96 well

Specifications

About this item

A Fluor-de-Lys® fluorescent assay system. The SIRT1 Fluorescent Activity Assay/Drug Discovery Kit is a complete assay system designed to measure the lysyl deacetylase activity of the recombinant human SIRT1 included in the kit.

  • Useful for inhibitor screening or characterizing enzyme kinetics
  • Includes optimal substrate selected from a panel of acetylated sites in p53 and histones

The Fluor-de-Lys® SIRT1 assay is based on the Fluor de Lys® SIRT1 Substrate and Fluor de Lys® Developer II combination. The assay procedure has two steps. First, the Fluor de Lys® SIRT1 Substrate, which contains a peptide comprising amino acids 379-382 of human p53 (Arg-His-Lys-Lys(Ac)). Deacetylation of the substrate sensitizes the substrate so that, in the second step, treatment with the Fluor de Lys®Developer II produces a fluorophore.

Delivery: Kit includes SIRT1 (Sirtuin 1, hSir2SIRT1) (human, recombinant), FLUOR DE LYS® SIRT1 Deacetylase Substrate, 5X FLUOR DE LYS® Developer II Concentrate, NAD (Sirtuin Substrate), Nicotinamide (Sirtuin Inhibitor), Resveratrol (Sirtuin Activator), Suramin sodium (Sirtuin Inhibitor), FLUOR DE LYS® Deacetylated Standard, Sirtuin Assay Buffer, Half volume microplates.