Order Entry
Hungary
ContactUsLinkComponent
Matrix Metalloproteinase-10 (MMP-10) fluorometric drug discovery kit
Matrix Metalloproteinase-10 (MMP-10) fluorometric drug discovery kit
Catalog # ENZOBMLAK4190001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Matrix Metalloproteinase-10 (MMP-10) fluorometric drug discovery kit
Catalog # ENZOBMLAK4190001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Description:
    Matrix Metalloproteinase-10 (MMP-10) fluorometric drug discovery kit
  • Seq:
    02
  • Storage conditions:
    −80 °C
  • Application:
    Colorimetric detection, HTS activity assay
  • Size:
    1 kit

Specifications

About this item

A QuantiZyme™ Assay System. The MMP-10 Fluorometric (also known as fluorimetric) Drug Discovery Kit is a complete assay sytem designed to screen for inhibitors of matrix metalloproteinase-10 (MMP-10, stromelysin-2) using a quenched fluorogenic peptide: OmniMMP™ fluorogenic substrate Mca-Pro-Leu-Gly-Leu-Dpa-Ala-Arg-NH. Mca fluorescence is quenched by the Dpa group until cleavage by MMPs at the Gly-Leu bond separates the two moieties.

Delivery: Kit contains MMP-10 Enzyme (human) (recombinant), Substrate, Calibration Standard, Prototypic Control Inhibitor (NNGH3), Assay Buffer, 96-well White Microplate and a detailed instruction booklet.