Order Entry
Hungary
ContactUsLinkComponent
Matrix Metalloproteinase-2 (MMP-2) fluorometric drug discovery kit
Matrix Metalloproteinase-2 (MMP-2) fluorometric drug discovery kit
Catalog # ENZOBMLAK4090001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Matrix Metalloproteinase-2 (MMP-2) fluorometric drug discovery kit
Catalog # ENZOBMLAK4090001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Description:
    Matrix Metalloproteinase-2 (MMP-2) fluorometric drug discovery kit
  • Seq:
    02
  • Storage conditions:
    −80 °C
  • Application:
    Colorimetric detection, HTS activity assay
  • Size:
    1 kit

Specifications

About this item

A Quantizyme™ Assay System. The MMP-2 Fluorometric (also known as fluorimetric) Drug Discovery Kit is a complete assay system designed to screen inhibitors of matrix metalloproteinase-2 (MMP-2, gelatinase A) using a quenched fluorogenic peptide: OmniMMP™ fluorogenic substrate Mca-Pro-Leu-Gly-Leu-Dpa-Ala-Arg-NH. Mca fluorescence is quenched by the Dpa group until cleavage by MMPs at the Gly-Leu bond separates the two moieties. The assays are performed in a convenient 96-well microplate format. The kit is useful to screen inhibitors of MMP-2, a potential therapeutic target.

Delivery: Kit contains MMP-2 enzyme (human) (recombinant), substrate, calibration standard, inhibitor, assay buffer and 96-well white microplate.