Order Entry
Hungary
ContactUsLinkComponent
Glutathione peroxidase activity kit
Glutathione peroxidase activity kit
Catalog # ENZOADI900158
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Glutathione peroxidase activity kit
Catalog # ENZOADI900158
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    Glutathione peroxidase activity kit
  • Seq:
    02
  • Size:
    96 wells
  • Tests per kit:
    5×96 wells
Specifications
About this item
The Glutathione Peroxidase activity kit is a convenient 96-well plate based assay for the higher throughput analysis of glutathione peroxidase activity in a variety of sample matrices in just 30 minutes.
For the measurement of glutathione peroxidase in cell lysates, plasma, and tissue from any species.
Ordering information: Contents: Microtiter plate, Assay buffer concentrate, Reaction mix, Glutathione reductase, GSSG