Proinsulin ELISA Kit, Grow Cells
Catalog # ENZOENZKIT1490001
Gyártó: ENZO LIFE SCIENCES
CAS szám:
Specifikációk
- Pk:1 KIT
- Description:Proinsulin ELISA kit, grow cells
- Seq:01
- Sensitivity:0,17 pM (1,56 - 50 pM)
Specifikációk
Termék információk
Ultra-sensitive Proinsulin ELISA kit enabling detection of proinsulin in human serum and plasma samples in just 3 hours.
- Ultra-sensitive measurement of proinsulin, detecting as little as 0,17 pM
- Negligible cross reactivity with insulin
- High throughput format with results in three hours for up to 40 samples in duplicate
- Fully quantitative results that surpass semi-quantitative Western blot analysis
For the quantitative determination of proinsulin in human serum and plasma samples.The Proinsulin ELISA kit is a colorimetric immunometric immunoassay kit with results in three hours.
Ordering information: Contains Microtiter plate, Assay Buffer 13, Standard, Wash Buffer Concentrate, Antibody, Antibody Diluent, Conjugate, Substrate, and Stop Solution.