Order Entry
Hungary
ContactUsLinkComponent
Adiponectin (human), ELISA kit
Adiponectin (human), ELISA kit
Catalog # ENZOALX850377KI01
Gyártó:  ENZO LIFE SCIENCES
CAS szám:  
Adiponectin (human), ELISA kit
Catalog # ENZOALX850377KI01
Gyártó:  ENZO LIFE SCIENCES
CAS szám:  

Specifikációk

  • Pk:
    1 KIT
  • Primary antibody reactivity:
    Human
  • Target protein:
    Adiponectin
  • Description:
    Adiponectin (human), ELISA kit
  • Seq:
    02
  • Sensitivity:
    0,156 ng/mL (CSF, urine); 0,47 ng/ml (plasma, serum) (range 1 to 150 ng/ml)

Specifikációk

Termék információk

Rapid, highly sensitive ELISA kit to quantify adiponectin for metabolic, diabetes, and obesity research.

For quantitative determination of human adiponectin in cerebrospinal fluid, plasma, serum, or urine.

The Adiponectin (human), ELISA kit is a colorimetric, immunometric immunoassay kit with results in < 3 hours.

Ordering information: Contains Microtiter Plate, Conjugate, Standards, Quality Controls, Dilution Buffer Concentrate, Substrate Solution, Stop Solution, Wash Buffer Concentrate.