Anti-RAP1A Rabbit Polyclonal Antibody
Catalog # ENZOADIKAPGP125E
Supplier: Enzo Life Sciences
Specifications
- Antibody type:Primary
- Antigen name:RAP1A, Member Of RAS Oncogene Family
- Antigen symbol:RAP1A
- Clonality:Polyclonal
- Host:Rabbit
- Western blot:Yes
- Seq:02
- Antigen synonyms:LRPAP1|KREV1|A2MRAP|RAP1|Low density lipoprotein receptor-related protein-associated protein 1|Alpha-2-macroglobulin receptor-associated protein|KREV-1|SMGP21
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Rap1, which is a member of the Ras family of GTP-binding proteins, cycles between an active GTP-bound and an unactive GDP-bound form that is mediated by GTPase activating protein (GAP). Rap1 is proposed to regulate Ras-mediated signaling and may also be involved in the regulation of Integrin-mediated cell adhesion although the mechanism of regulation is not known.
- For Western Blot
Recognizes human, mouse, rat, bovine, dog, chicken, guinea pig, hamster, monkey, pig, rabbit, sheep and Xenopus Rap1. Detects a band of ~25kDa by Western blot.
Type: Primary
Antigen: RAP1A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human